accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 128544-05-8

IUPAC Name: 2'-(trifluoromethanesulfonyloxy)-[1,1'-binaphthalen]-2-yl trifluoromethanesulfonate
Synonyms: 2'-(trifluoromethanesulfonyloxy)-[1,1'-binaphthalen]-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 550.44
Molecular Formula: C22H12F6O6S2
InChi Key: OYJLCOSEYYZULE-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC=C2C=CC=CC2=C1C1=C(OS(=O)(=O)C(F)(F)F)C=CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results