accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCRuCCCCC

CAS RN 1287-13-4

IUPAC Name: ruthenocene
Synonyms: ruthenocene
Molecular Weight (g/mol): 226.22
Molecular Formula: C10H5Ru
InChi Key: —
SMILES: c1cc[c-]c1.[C-]12[C-]3[C-]4[C-]5[C-]1[Ru]2345
Quantity 
  • (3)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)