Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenocene, TCI America™
SDP

Catalog No. R00781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R00781G Supplier TCI America Supplier No. R00781G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1287-13-4
Molecular Formula C10H10Ru
MDL Number MFCD00001442
Synonym Bis(cyclopentadienyl)ruthenium(II)

Specifications

Melting Point 202°C
Color Yellow
Quantity 1 g
Formula Weight 231.26
Physical Form Crystalline Powder
Chemical Name or Material Ruthenocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.