CAS RN 129-00-0
Pyrene, 98%
CAS: 129-00-0 Molecular Formula: C16H10 Molecular Weight (g/mol): 202.25 MDL Number: MFCD00004136 InChI Key: BBEAQIROQSPTKN-UHFFFAOYSA-N Synonym: benzo def phenanthrene,pyren,beta-pyrene,.beta.-pyrene,pyren german,unii-9e0t7wfw93,ccris 1256,pyrene def phenanthrene,coal tar pitch volatiles:pyrene PubChem CID: 31423 ChEBI: CHEBI:39106 IUPAC Name: pyrene SMILES: C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
Pyrene, 98%
CAS: 129-00-0 Molecular Formula: C16H10 Molecular Weight (g/mol): 202.256 MDL Number: MFCD00004136 InChI Key: BBEAQIROQSPTKN-UHFFFAOYSA-N Synonym: benzo def phenanthrene,pyren,beta-pyrene,.beta.-pyrene,pyren german,unii-9e0t7wfw93,ccris 1256,pyrene def phenanthrene,coal tar pitch volatiles:pyrene PubChem CID: 31423 ChEBI: CHEBI:39106 IUPAC Name: pyrene SMILES: C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
Pyrene, MilliporeSigma™ Supelco™
Pyrene belongs to polycyclic aromatic hydrocarbon group. These are organic compounds which have two or more fused benzene rings in linear, angular and cluster arrangements. Pyrene is a four ring PAH.
Pyrene 97.0+%, TCI America™
CAS: 129-00-0 Molecular Formula: C16H10 Molecular Weight (g/mol): 202.256 MDL Number: MFCD00004136 InChI Key: BBEAQIROQSPTKN-UHFFFAOYSA-N Synonym: benzo def phenanthrene,pyren,beta-pyrene,.beta.-pyrene,pyren german,unii-9e0t7wfw93,ccris 1256,pyrene def phenanthrene,coal tar pitch volatiles:pyrene PubChem CID: 31423 ChEBI: CHEBI:39106 IUPAC Name: pyrene SMILES: C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
Pyrene (purified by sublimation) 98.0+%, TCI America™
CAS: 129-00-0 Molecular Formula: C16H10 Molecular Weight (g/mol): 202.256 MDL Number: MFCD00004136 InChI Key: BBEAQIROQSPTKN-UHFFFAOYSA-N Synonym: benzo def phenanthrene,pyren,beta-pyrene,.beta.-pyrene,pyren german,unii-9e0t7wfw93,ccris 1256,pyrene def phenanthrene,coal tar pitch volatiles:pyrene PubChem CID: 31423 ChEBI: CHEBI:39106 IUPAC Name: pyrene SMILES: C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
Sigma Aldrich Pyrene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Pyrene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Pyrene Solution, 100 μg/mL, Methylene Chloride, 1 mL
CAS: 129-00-0 Molecular Formula: C16H10 Molecular Weight (g/mol): 202.256 InChI Key: BBEAQIROQSPTKN-UHFFFAOYSA-N PubChem CID: 31423 ChEBI: CHEBI:39106 IUPAC Name: pyrene SMILES: C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
PAH - Sediment 1, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Pyrene, SPEX CertiPrep™
CAS: 129-00-0 Molecular Formula: C16H10 Molecular Weight (g/mol): 202.256 InChI Key: BBEAQIROQSPTKN-UHFFFAOYSA-N PubChem CID: 31423 ChEBI: CHEBI:39106 IUPAC Name: pyrene SMILES: C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2