Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrene, MilliporeSigma™ Supelco™

Catalog No. 111019998 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9998 5000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9998 Supplier MilliporeSigma Supelco Supplier No. 48570
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrene belongs to polycyclic aromatic hydrocarbon group. These are organic compounds which have two or more fused benzene rings in linear, angular and cluster arrangements. Pyrene is a four ring PAH.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 129-00-0
Flash Point >200°C
Physical Form Neat
Linear Formula C16H10
MDL Number MFCD00004136
Melting Point 145°C to 148°C
Synonym Benzo[def]phenanthrene
Type Organic Pollutant Standard
Quantity 5000 mg
Formula Weight 202.25
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.