accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129034-70-4

IUPAC Name: 3-(propan-2-yloxy)-4-(tributylstannyl)cyclobut-3-ene-1,2-dione
Synonyms: 3-isopropoxy-4-(tributylstannyl)cyclobut-3-ene-1,2-dione
Molecular Weight (g/mol): 429.19
Molecular Formula: C19H34O3Sn
InChi Key: AMVLEBHYELFWJT-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=C(OC(C)C)C(=O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results