accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeCH3CH3

CAS RN 1291-47-0

IUPAC Name: 1,1'-Dimethylferrocene
Synonyms: —
Molecular Weight (g/mol): 214.09
Molecular Formula: C12H14Fe
InChi Key: —
SMILES: [Fe].Cc1cccc1.Cc1cccc1
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)