Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Dimethylferrocene 93.0+%, TCI America™
SDP

Catalog No. D12735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12735G 5 g
D127325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12735G Supplier TCI America Supplier No. D12735G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1291-47-0
Molecular Formula C12H14Fe
Molecular Weight (g/mol) 214.09
MDL Number MFCD00001425
InChI Key SNGSYGKLJUGRID-UHFFFAOYSA-N
IUPAC Name 1,1'-Dimethylferrocene
SMILES [Fe].Cc1cccc1.Cc1cccc1

Specifications

Melting Point 40°C
Color Red-Yellow
Quantity 5 g
Formula Weight 214.09
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Dimethylferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.