accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129103-69-1

IUPAC Name: 2-bromo-4,5-dimethoxyphenol
Synonyms: 2-bromo-4,5-dimethoxyphenol
Molecular Weight (g/mol): 233.06
Molecular Formula: C8H9BrO3
InChi Key: YGMAGXHPUIGKFF-UHFFFAOYSA-N
SMILES: COC1=CC(O)=C(Br)C=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results