CAS RN 129203-60-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Iberiotoxin, Buthus tamulus, Recombinant, E. coli
Recombinant Buthus tamulus iberiotoxin expressed in E.