Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Iberiotoxin, Buthus tamulus, Recombinant, E. coli

Catalog No. 40100210UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-100-210UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-100-210UG Supplier MilliporeSigma™ Supplier No. 40100210UG
Add to Cart
Add to Cart

Recombinant Buthus tamulus iberiotoxin expressed in E.

Peptide Sequence: Pyr-Phe-Thr-Asp-Val-Asp-Cys7-Ser-Val-Ser-Lys-Glu-Cys13-Trp-Ser-Val-Cys17-Lys-Asp-Leu-Phe-Gly-Val-Asp-Arg-Gly-Lys-Cys28-Met-Gly-Lys-Lys-Cys33-Arg-Cys35-Tyr-Gln-OH (disulfide bonds: 7 →28; 13 → 35; 17 → 33)
TRUSTED_SUSTAINABILITY

Specifications

Description 98% by HPLC
Target Proteins High conductance Ca2+-activated K+ channels
CAS 129203-60-7
Formula Weight 4231g/mol
Solubility 100 mM NaCl, 10 mM Tris-HCl, pH 7.5 containing 0.1% BSA
Form Lyophilized
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.