CAS RN 129211-47-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Azoniaspiro[4.4]nonane Tetrafluoroborate 98.0+%, TCI America™
CAS: 129211-47-8 Molecular Formula: C8H16BF4N