Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Azoniaspiro[4.4]nonane Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. A31035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31035G 5 g
A31031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31035G Supplier TCI America Supplier No. A31035G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 129211-47-8
Molecular Formula C8H16BF4N

Specifications

Melting Point 190°C
Color White
Quantity 5 g
UN Number 1759
Formula Weight 213.03
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Azoniaspiro[4.4]nonane Tetrafluoroborate
EINECSNumber (5)-6908
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.