accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129223-22-9

IUPAC Name: 1-(1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-2-carbonyl)pyrrolidine-2-carboxylic acid
Synonyms: 1-{1-[(9H-fluoren-9-ylmethoxy)carbonyl]pyrrolidine-2-carbonyl}pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 434.49
Molecular Formula: C25H26N2O5
InChi Key: VRAQFWSWKRNOGU-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCN1C(=O)C1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)