Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-prolyl-L-proline, 95%, Thermo Scientific Chemicals

Catalog No. AAH6622806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6622806 5 g
AAH6622803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6622806 Supplier Thermo Scientific Chemicals Supplier No. H6622806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129223-22-9
Molecular Formula C25H26N2O5
Molecular Weight (g/mol) 434.492
MDL Number MFCD00237673
InChI Key VRAQFWSWKRNOGU-VXKWHMMOSA-N
Synonym fmoc-pro-pro-oh, s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid, l-proline, 1-9h-fluoren-9-ylmethoxy carbonyl-l-prolyl, 2s-1-2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid, s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid, fmoc-pro-pro, fmoc-prolinyl-proline, fmoc-prolylproline-oh, fmoc-l-pro-l-pro-oh, fmoc-prolinyl-proline s,s
PubChem CID 11133845
IUPAC Name (2S)-1-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carbonyl]pyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=O)C2CCCN2C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O

Specifications

Quantity 5 g
Formula Weight 434.49
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Fmoc-L-prolyl-L-proline
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.