CAS RN 129298-91-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ TNP-470
TNP-470, CAS 129298-91-5, is a cell-permeable Fumagillin (Cat. No. 344845) analog that specifically inhibits MetAP2 methionine aminopeptidase activity (IC50 = 1nM for human MetAP2).