accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129394-11-2

IUPAC Name: 1-(pyridin-2-yl)piperazine hydrochloride
Synonyms: pyridinylpiperazine hydrochloride
Molecular Weight (g/mol): 199.68
Molecular Formula: C9H14ClN3
InChi Key: XQVSONPKJXNWJO-UHFFFAOYSA-N
SMILES: Cl.C1CN(CCN1)C1=CC=CC=N1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results