Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Pyridyl)piperazine Monohydrochloride, TCI America™
SDP

Catalog No. P133725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P133725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P133725G Supplier TCI America Supplier No. P133725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129394-11-2
Molecular Formula C9H14ClN3
Molecular Weight (g/mol) 199.682
MDL Number MFCD01861295
InChI Key XQVSONPKJXNWJO-UHFFFAOYSA-N
Synonym 2-(1-Piperazinyl)pyridine Monohydrochloride
PubChem CID 22097317
IUPAC Name 1-pyridin-2-ylpiperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=CC=N2.Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 199.68
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Pyridyl)piperazine Monohydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.