accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONH3CBr

CAS RN 129477-21-0

IUPAC Name: ethyl 5-bromo-2-methylpyridine-3-carboxylate
Synonyms: ethyl 5-bromo-2-methylpyridine-3-carboxylate
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: QVKPPFGWQMCQCI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(Br)=CN=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results