Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-bromo-2-methylnicotinate, 97%, Thermo Scientific™

Catalog No. AAH6138503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6138503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6138503 Supplier Thermo Scientific Chemicals Supplier No. H6138503

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 5-bromo-2-methylnicotinate is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129477-21-0
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD10698571
InChI Key QVKPPFGWQMCQCI-UHFFFAOYSA-N
PubChem CID 15077476
IUPAC Name ethyl 5-bromo-2-methylpyridine-3-carboxylate
SMILES CCOC(=O)C1=CC(=CN=C1C)Br

Specifications

Color Yellow
Quantity 1 g
Synonym ethyl 5-bromo-2-methylnicotinate, 5-bromo-2-methyl-nicotinic acid ethyl ester, 5-bromo-2-methyl-3-pyridinecarboxylic acid ethyl ester, 5-bromo-2-methylnicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 5-bromo-2-methyl-, ethyl ester, ethyl5-bromo-2-methylnicotinate, ethyl 5-bromo-2-methyl-pyridine-3-carboxylate
Solubility Information Slightly soluble in water.
Formula Weight 244.09
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 5-bromo-2-methylnicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.