accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129486-91-5

IUPAC Name: 2-methyl-1H-imidazole-5-carbothioamide
Synonyms: 2-methyl-3H-imidazole-4-carbothioamide
Molecular Weight (g/mol): 141.19
Molecular Formula: C5H7N3S
InChi Key: MNYPQSNAWPZXRV-UHFFFAOYSA-N
SMILES: CC1=NC=C(N1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results