Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1H-imidazole-4-carbothioamide, 97%, Thermo Scientific™

Catalog No. SB02117DE
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB02117DE 5 g
SB02117DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB02117DE Supplier Thermo Scientific Chemicals Supplier No. SB02117DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129486-91-5
Molecular Formula C5H7N3S
Molecular Weight (g/mol) 141.192
MDL Number MFCD03659724
InChI Key MNYPQSNAWPZXRV-UHFFFAOYSA-N
Synonym 2-methyl-1h-imidazole-4-carbothioamide, 1h-imidazole-5-carbothioamide,2-methyl, 1h-imidazole-5-carbothioamide, 2-methyl, acmc-1c27g, amino 2-methylimidazol-4-yl methane-1-thione, 2-methyl-3h-imidazole-4-carbothioamide
PubChem CID 2796714
IUPAC Name 2-methyl-1H-imidazole-5-carbothioamide
SMILES CC1=NC=C(N1)C(=S)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 141.2
Percent Purity 97%
Chemical Name or Material 2-Methyl-1H-imidazole-4-carbothioamide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.