accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129619-37-0

IUPAC Name: (3R,6R)-octane-3,6-diol
Synonyms: (3R,6R)-octane-3,6-diol
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: BCKOQWWRTRBSGR-HTQZYQBOSA-N
SMILES: CC[C@@H](O)CC[C@H](O)CC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results