Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R,6R)-3,6-Octanediol 96.0+%, TCI America™
SDP

Catalog No. O0293500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0293500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O0293500MG Supplier TCI America Supplier No. O0293500MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 129619-37-0
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD01631087
InChI Key BCKOQWWRTRBSGR-HTQZYQBOSA-N
Synonym 3r,6r-3,6-octanediol, 3r,6r-octane-3,6-diol, 3,6-octanediol, 3r,6r, 3r,3r-3,6-octadiol, 3r,6r-3,6-dihydroxyoctane, 3r,6r---3,6-octanediol
PubChem CID 2734541
IUPAC Name (3R,6R)-octane-3,6-diol
SMILES CC[C@@H](O)CC[C@H](O)CC

Specifications

Color White
Quantity 500 mg
Formula Weight 146.23
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (3R,6R)-3,6-Octanediol
EINECSNumber (2)-0240
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.