accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClFFF

CAS RN 129825-11-2

IUPAC Name: 1-[4-chloro-3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[4-chloro-3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 222.59
Molecular Formula: C9H6ClF3O
InChi Key: UYNMUXTXDHJBEN-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Cl)C(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)