Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chloro-3'-(trifluoromethyl)acetophenone 98.0+%, TCI America™
SDP

Catalog No. C28215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C28215G Supplier TCI America Supplier No. C28215G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129825-11-2
Molecular Formula C9H6ClF3O
Molecular Weight (g/mol) 222.591
MDL Number MFCD03094146
InChI Key UYNMUXTXDHJBEN-UHFFFAOYSA-N
Synonym 4'-chloro-3'-trifluoromethyl acetophenone, 4-chloro-3-trifluoromethyl acetophenone, 1-4-chloro-3-trifluoromethyl phenyl ethanone, 1-4-chloro-3-trifluoromethyl phenyl ethan-1-one, 3'-trifluoromethyl-4'-chloroacetophenone, 5-acetyl-2-chlorobenzotrifluoride, ethanone,1-4-chloro-3-trifluoromethyl phenyl, ethanone, 1-4-chloro-3-trifluoromethyl phenyl, 3-trifluoromethyl-4-chloroacetophenone
PubChem CID 2773851
IUPAC Name 1-[4-chloro-3-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC(=C(C=C1)Cl)C(F)(F)F

Specifications

Melting Point 62°C
Color White
Quantity 5 g
Formula Weight 222.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-3′-(trifluoromethyl)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.