accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOCH3OHOZn

CAS RN 129918-15-6

IUPAC Name: bis(2-methoxyethan-1-ol) zinc
Synonyms: bis(2-methoxyethanol) zinc
Molecular Weight (g/mol): 217.57
Molecular Formula: C6H16O4Zn
InChi Key: SZCCOQAZCQRATH-UHFFFAOYSA-N
SMILES: [Zn].COCCO.COCCO
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)