Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc 2-methoxyethoxide, 5% w/v in 2-methoxyethanol

Catalog No. AA43903AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43903AE 100 mL
AA43903AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43903AE Supplier Thermo Scientific Chemicals Supplier No. 043903AE
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 129918-15-6 | C6H16O4Zn | 217.57 g/mol

Zinc 2-methoxyethoxide is used as catalyst component in oxirane polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129918-15-6
Molecular Formula C6H16O4Zn
Molecular Weight (g/mol) 217.57
MDL Number MFCD00156529
InChI Key SZCCOQAZCQRATH-UHFFFAOYSA-N

Specifications

Color Yellow
Density 0.96 g/mL
Boiling Point 124°C
Flash Point 40°C (104°F)
Linear Formula Zn(OCH2CH2OCH3)2
Quantity 100 mL
UN Number UN1188
Sensitivity Moisture sensitive
Solubility Information Soluble in methoxyethanol and pyridine. Miscible with alcohols,ketones,esters and water.
Formula Weight 215.56
Physical Form Liquid
Chemical Name or Material Zinc 2-methoxyethoxide
Show More Show Less
Hazard Category H226-H302+H312+H332-H314-H335-H360FD-H370-H373
Hazard Statement GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.