accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3COOSOOO

CAS RN 130-37-0

IUPAC Name: sodium 2-methyl-1,4-dioxo-1,2,3,4-tetrahydronaphthalene-2-sulfonate
Synonyms: menadione sodium bisulfite vitamin K3 sodium bisulfite menaphthone sodium bisulphite bisulfito sodico de menadiona menadioni natrii bisulfis sodium menadione bisulfite menadione sodium sulfonate bisulfite sodique de menadione menaphthone sodium bisulfite natrium menadionsulfonicum
Molecular Weight (g/mol): 276.24
Molecular Formula: C11H9NaO5S
InChi Key: XDPFHGWVCTXHDX-UHFFFAOYNA-M
SMILES: [Na+].CC1(CC(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results