Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Menadione Sodium Bisulfite Trihydrate, MP Biomedicals™

Catalog No. 0210226091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-102-26091 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-102-26091 Supplier MP Biomedicals, Inc Supplier No. 0210226091
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Menadione Sodium Bisulfite Trihydrate is a water-soluble form of menadione which belongs to the Vitamin K class of compounds.

  • Menadione is a prothrombogenic compound and is used as a model quinone in cell culture and in vivo investigations.
  • Menadione has been shown to affect gap-junctional intercellular communication by mediation of tyrosine phosphorylation.
  • Menadione has demonstrated cytotoxic activity against a variety of cell lines and can induce apoptosis in cultured cells, such as osteoclasts and osteoblasts, via elevation of peroxide and superoxide radical levels.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-37-0
Molecular Formula C11H9NaO5S
Molecular Weight (g/mol) 276.24
InChI Key XDPFHGWVCTXHDX-UHFFFAOYNA-M
IUPAC Name sodium 2-methyl-1,4-dioxo-1,2,3,4-tetrahydronaphthalene-2-sulfonate
SMILES [Na+].CC1(CC(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O

Specifications

Quantity 1 kg
Percent Purity >95%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.