accessibility menu, dialog, popup

UserName

Viewing profile as user:
BaOSOOHNOSOOHN

CAS RN 1300-92-1

IUPAC Name: barium(2+) bis(2-(phenylamino)benzene-1-sulfonate)
Synonyms: barium(2+) bis(2-(phenylamino)benzenesulfonate)
Molecular Weight (g/mol): 633.88
Molecular Formula: C24H20BaN2O6S2
InChi Key: JWTHKGUPPCYKNI-UHFFFAOYSA-L
SMILES: [Ba++].[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=CC=C1.[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=CC=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results