Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Diphenylamine Sulfonate, Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCB009025F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Pouch
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCB009025F 25 g Pouch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCB009025F Supplier Ricca Chemical Company Supplier No. RDCB009025F1

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1300-92-1
Molecular Weight (g/mol) 633.88 g/mol

Specifications

Color White to Gray
CAS Min % 98.0
CAS Max % 100.0
Physical Form Solid
Quantity 25 g
Grade Reagent
Packaging Pouch
Chemical Name or Material Barium Diphenylamine Sulfonate
ShelfLife 60 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.