accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 130018-87-0

IUPAC Name: 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dihydrochloride
Synonyms: cetirizine dihydrochloride
Molecular Weight (g/mol): 461.81
Molecular Formula: C21H27Cl3N2O3
InChi Key: PGLIUCLTXOYQMV-UHFFFAOYNA-N
SMILES: Cl.Cl.OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results