CAS RN 13005-35-1
IUPAC Name:
copper(2+) bis(2,3,4,5,6-pentahydroxyhexanoate)
Synonyms:
copper(2+) bis(2,3,4,5,6-pentahydroxyhexanoate)
Molecular Weight (g/mol):
453.84
Molecular Formula:
C12H22CuO14
InChi Key:
OCUCCJIRFHNWBP-UHFFFAOYNA-L
SMILES:
[Cu++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O
1
–
1
of
1
results
1
MP Biomedicals, Inc Copper Gluconate, MP Biomedicals
CAS: 13005-35-1 Molecular Formula: C12H22CuO14 Molecular Weight (g/mol): 453.84 InChI Key: OCUCCJIRFHNWBP-UHFFFAOYSA-L Synonym: copper d-gluconate,copper ii gluconate,bis d-gluconato copper,cupric gluconate monohydrate,chelates of copper gluconate,copper, bis d-gluconato,copper d-gluconate 1:2,ccris 3652,hsdb 261,bis d-gluconato-o1,o2 copper PubChem CID: 93070 IUPAC Name: copper;2,3,4,5,6-pentahydroxyhexanoate SMILES: C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Cu+2]