Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Gluconate, MP Biomedicals

Catalog No. ICN203919 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203919 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203919 Supplier MP Biomedicals, Inc Supplier No. 0520391905

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13005-35-1
Molecular Formula C12H22CuO14
Molecular Weight (g/mol) 453.84
InChI Key OCUCCJIRFHNWBP-UHFFFAOYSA-L
Synonym copper d-gluconate, copper ii gluconate, bis d-gluconato copper, cupric gluconate monohydrate, chelates of copper gluconate, copper, bis d-gluconato, copper d-gluconate 1:2, ccris 3652, hsdb 261, bis d-gluconato-o1,o2 copper
PubChem CID 93070
IUPAC Name copper;2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Cu+2]

Specifications

Quantity 100 g
Formula Weight 453.85
Chemical Name or Material Copper Gluconate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.