CAS RN 13013-02-0
IUPAC Name:
methyl 4-nitrobutanoate
Synonyms:
methyl 4-nitrobutanoate
Molecular Weight (g/mol):
147.13
Molecular Formula:
C5H9NO4
InChi Key:
UBSPKGKFFQKZJB-UHFFFAOYSA-N
SMILES:
COC(=O)CCC[N+]([O-])=O
1
–
1
of
1
results
1
Sigma Aldrich Methyl 4-nitrobutyrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.