accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClCl

CAS RN 13014-18-1

IUPAC Name: 2,4-dichloro-1-(trichloromethyl)benzene
Synonyms: 2,4-dichloro-1-(trichloromethyl)benzene
Molecular Weight (g/mol): 264.35
Molecular Formula: C7H3Cl5
InChi Key: KZSNBJMYJWDVTK-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C(Cl)=C1)C(Cl)(Cl)Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results