Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzotrichloride 99.0+%, TCI America™
SDP

Catalog No. D042825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D042825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D042825G Supplier TCI America Supplier No. D042825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 13014-18-1
Melting Point 49°C
Color White-Yellow
Boiling Point 159°C
Molecular Formula C7H3Cl5
Quantity 25 g
MDL Number MFCD00018818
Synonym alpha,alpha,alpha,2,4-Pentachlorotoluene
InChI Key KZSNBJMYJWDVTK-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)Cl)C(Cl)(Cl)Cl
IUPAC Name 2,4-dichloro-1-(trichloromethyl)benzene
Molecular Weight (g/mol) 264.351
PubChem CID 25607
Formula Weight 264.35
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorobenzotrichloride
Show More Show Less
EINECSNumber (3)-0093
RTECSNumber CZ5475020
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.