accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH3CCH3CH3NH2H2NCH3HClHCl

CAS RN 1301706-47-7

IUPAC Name: 1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine dihydrochloride
Synonyms: 1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine dihydrochloride
Molecular Weight (g/mol): 369.37
Molecular Formula: C20H30Cl2N2
InChi Key: HSFKFAWYBTVLDG-UHFFFAOYNA-N
SMILES: Cl.Cl.CC1=CC(C)=C(C(N)C(N)C2=C(C)C=C(C)C=C2C)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)