Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-1,2-Bis(2,4,6-trimethylphenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%, Thermo Scientific™

Catalog No. AAH25988MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25988MD 250 mg
AAH2598803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25988MD Supplier Thermo Scientific Chemicals Supplier No. H25988MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1301706-47-7
Molecular Formula C20H30Cl2N2
Molecular Weight (g/mol) 369.374
MDL Number MFCD06797063
InChI Key HSFKFAWYBTVLDG-WUMQWIPTSA-N
Synonym r,r-bis-mesityl ethylenediamine dihydrochloride, r,r-1,2-bis 2,4,6-trimethylphenyl-1,2-ethanediamine dihydrochloride, 1r,2r-1,2-bis 2,4,6-trimethylphenyl ethane-1,2-diamine dihydrochloride, r,r-1,2-bis 2,4,6-trimethylphenyl-1,2-ethanediamine dihydrochloride, ee
PubChem CID 45925672
IUPAC Name (1R,2R)-1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine;dihydrochloride
SMILES CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C.Cl.Cl

Specifications

Quantity 250 mg
Formula Weight 369.38
Percent Purity 95%
Assay ee 99%
Chemical Name or Material (R,R)-1,2-Bis(2,4,6-trimethylphenyl)-1,2-ethanediamine dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.