accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHNH2O

CAS RN 13027-88-8

IUPAC Name: 2-hydroxy-2-methylpropanamide
Synonyms: 2-hydroxy-2-methylpropanamide
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: DRYMMXUBDRJPDS-UHFFFAOYSA-N
SMILES: CC(C)(O)C(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results