Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyisobutyramide 98.0+%, TCI America™
SDP

Catalog No. H118125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H118125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H118125G Supplier TCI America Supplier No. H118125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13027-88-8
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD08063365
InChI Key DRYMMXUBDRJPDS-UHFFFAOYSA-N
Synonym 2-Hydroxy-2-methylpropionamide
PubChem CID 83059
IUPAC Name 2-hydroxy-2-methylpropanamide
SMILES CC(C)(C(=O)N)O

Specifications

Melting Point 93°C
Color White
Quantity 25 g
Formula Weight 103.12
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyisobutyramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.