accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 13029-08-8

IUPAC Name: 2,2'-dichloro-1,1'-biphenyl
Synonyms: 2,2'-dichlorobiphenyl
Molecular Weight (g/mol): 223.1
Molecular Formula: C12H8Cl2
InChi Key: JAYCNKDKIKZTAF-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results