accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1303-33-9

IUPAC Name: diarsenic(3+) trisulfanediide
Synonyms: arsenic sulfide
Molecular Weight (g/mol): 246.02
Molecular Formula: As2S3
InChi Key: CUGMJFZCCDSABL-UHFFFAOYSA-N
SMILES: [S--].[S--].[S--].[As+3].[As+3]
Physical Form 
  • (2)
  • (2)
  • (5)
Quantity 
  • (1)
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (2)
  • (5)
  • (1)
  • (4)
  • (4)
  • (2)
  • (2)