Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic(III) sulfide, 99.9% (metals basis)

Catalog No. AA4334014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4334014 25 g
AA4334006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4334014 Supplier Thermo Scientific Chemicals Supplier No. 04334014
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1303-33-9 | As2S3 | 246.02 g/mol

For vacuum deposition.Arsenic(III) sulfide is used to make infrared-transmitting glass, pyrotechnics, pigments, linoleum, electrical semiconductors and photoconductors. It acts as a reducing agent and serves as a tanning agent. It is also used for depilating hides and veterinary medication. Further, it is used as an inorganic photoresist to fabricate 3-D nanostructures. In addition to this, it is used in the treatment for acute promyelocytic leukemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-33-9
Molecular Formula As2S3
Molecular Weight (g/mol) 246.02
MDL Number MFCD00003435
InChI Key CUGMJFZCCDSABL-UHFFFAOYSA-N
IUPAC Name diarsenic(3+) trisulfanediide
SMILES [S--].[S--].[S--].[As+3].[As+3]

Specifications

Boiling Point 707°C
Melting Point 300°C
Color Yellow
Physical Form Fused Pieces
Quantity 25 g
Odor Like rotten eggs
Assay Percent Range 99.99% (Metals basis)
Density 3.43
Solubility Information Insoluble in water.
Formula Weight 246.03
Chemical Name or Material Arsenic(III) sulfide
Show More Show Less
Hazard Category H301+H331-H350
Hazard Statement When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 215-117-4
RTECSNumber CG2638000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.