accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOCH3OOCH3OOCH3O

CAS RN 13035-61-5

IUPAC Name: [3,4,5-tris(acetyloxy)oxolan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)oxolan-2-yl]methyl acetate
Molecular Weight (g/mol): 318.28
Molecular Formula: C13H18O9
InChi Key: IHNHAHWGVLXCCI-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (2)
Quantity 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (4)