Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%

Catalog No. AC170121000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170121000 100 g Glass bottle
AC170120250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170121000 Supplier Thermo Scientific Chemicals Supplier No. 170121000
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 13035-61-5 | C13H18O9 | 318.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13035-61-5
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.27
MDL Number MFCD00005358
InChI Key IHNHAHWGVLXCCI-FDYHWXHSSA-N
Synonym beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
PubChem CID 83064
IUPAC Name [(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 79°C to 81°C
Color White
Ignition Residue 0.1% max.
Loss on Drying 1% max. (60°C, 2 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 100 g
Specific Rotation Condition −15.40° (20°C c=7,MeOH)
Specific Rotation -15.4
Formula Weight 318.27
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material β-D-Ribofuranose 1,2,3,5-tetraacetate
Show More Show Less
EINECSNumber 235-898-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.