accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNCS

CAS RN 13037-60-0

IUPAC Name: 1-bromo-2-isothiocyanatobenzene
Synonyms: 1-bromo-2-isothiocyanatobenzene
Molecular Weight (g/mol): 214.08
Molecular Formula: C7H4BrNS
InChi Key: PAFORXDSYWMYGP-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1N=C=S
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)