Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromophenyl Isothiocyanate 97.0+%, TCI America™
SDP

Catalog No. I05235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05235G Supplier TCI America Supplier No. I05235G

Chemical Identifiers

CAS 13037-60-0
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD00004799
InChI Key PAFORXDSYWMYGP-UHFFFAOYSA-N
Synonym 2-bromophenyl isothiocyanate, benzene,1-bromo-2-isothiocyanato, benzene, 1-bromo-2-isothiocyanato, 2-bromophenylisothiocyanate, benzene, bromoisothiocyanato, isothiocyanic acid 2-bromophenyl ester, acmc-1bbbv, o-bromophenyl isothiocyanate, 1-bromo-2-isothiocyanato-benzene, #
PubChem CID 611654
IUPAC Name 1-bromo-2-isothiocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=S)Br

Specifications

Color Yellow
Boiling Point 118°C
Quantity 5 g
UN Number 1760
Formula Weight 214.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromophenyl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.