accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13037-86-0

H3COOH

CAS RN 13037-86-0

IUPAC Name: 4-(heptyloxy)phenol
Synonyms: 4-heptyloxyphenol
Molecular Weight (g/mol): 208.3
Molecular Formula: C13H20O2
InChi Key: HZBABTUFXQLADL-UHFFFAOYSA-N
SMILES: CCCCCCCOC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)